Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: GLIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343419.1 | NP_001332581.1 | 3 - 4 | 1247 | 106 | |
NM_121576.2 | NP_568318.1 | 4 - 5 | 1195 | 106 | ||
GenBank mRNA | AY084802.1 | - | 4 - 5 | 890 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |