Search
Search

CSLA11| Gene Symbol: CSLA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343440.1 | NP_001330943.1 | 5 - 6 | 1229 | 67 | |
| NM_001343441.1 | NP_001330942.1 | 5 - 6 | 1229 | 67 | ||
| NM_001343442.1 | NP_001330941.1 | 5 - 6 | 1167 | 67 | ||
| NM_121624.4 | NP_197123.3 | 5 - 6 | 680 | 67 | ||
| GenBank mRNA | KJ138686.1 | - | 5 - 6 | 680 | 67 | |
Set Membership: |
|