Search
Search

GLN1;4| Gene Symbol: GLN1;4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343456.1 | NP_001331815.1 | 8 - 9 | 760 | 63 | |
| NM_121663.3 | NP_568335.1 | 8 - 9 | 894 | 63 | ||
| GenBank mRNA | AY059932.1 | - | 8 - 9 | 711 | 63 | |
| AY128749.1 | - | 8 - 9 | 668 | 63 | ||
Set Membership: |
|