Search
Search

DOT2| Gene Symbol: DOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343464.1 | NP_001318573.1 | 8 - 9 | 1383 | 65 | |
| NM_001343465.1 | NP_001330040.1 | 8 - 9 | 1380 | 65 | ||
| NM_121684.4 | NP_197180.1 | 8 - 9 | 1380 | 65 | ||
Set Membership: |
|