Search
Search

XPO1A| Gene Symbol: XPO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203395.1 | NP_001190324.1 | 26 - 27 | 2753 | 110 | |
| NM_121708.3 | NP_197204.1 | 27 - 28 | 2890 | 110 | ||
| GenBank mRNA | AY056126.1 | - | 27 - 28 | 2786 | 110 | |
| AY096633.1 | - | 27 - 28 | 2618 | 110 | ||
| Y18469.1 | - | 2713 | 110 | |||
Set Membership: |
|