Search
Search

SPL7| Gene Symbol: SPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203404.1 | NP_001190333.1 | 9 - 10 | 2232 | 111 | |
| NM_121888.3 | NP_197384.1 | 9 - 10 | 2327 | 111 | ||
| GenBank mRNA | AF367355.1 | - | 9 - 10 | 2168 | 111 | |
| AJ011612.1 | - | 9 - 10 | 2126 | 111 | ||
Set Membership: |
|