Search
Search

AT5G19070| Gene Symbol: AT5G19070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121912.3 | NP_197408.2 | 8 - 9 | 1043 | 67 | |
| GenBank mRNA | AK228918.1 | - | 8 - 9 | 864 | 67 | |
| BT014970.1 | - | 8 - 9 | 880 | 67 | ||
| BT020544.1 | - | 8 - 9 | 633 | 67 | ||
Set Membership: |
|