Search
Search

GSTF9| Gene Symbol: GSTF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084514.1 | NP_001077983.1 | 1 - 2 | 273 | 81 | |
| NM_128638.3 | NP_180643.1 | 1 - 2 | 388 | 81 | ||
| GenBank mRNA | AF372905.1 | - | 1 - 2 | 189 | 81 | |
| BT002679.1 | - | 1 - 2 | 138 | 81 | ||
Set Membership: |
|