Search
Search

GSTF9| Gene Symbol: GSTF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084514.1 | NP_001077983.1 | 2 - 3 | 329 | 66 | |
| NM_128638.3 | NP_180643.1 | 2 - 3 | 444 | 66 | ||
| GenBank mRNA | AF372905.1 | - | 2 - 3 | 245 | 66 | |
| BT002679.1 | - | 2 - 3 | 194 | 66 | ||
Set Membership: |
|