Search
Search

ARI7| Gene Symbol: ARI7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336334.1 | NP_001324416.1 | 3 - 4 | 799 | 73 | |
| NM_001336335.1 | NP_001324417.1 | 3 - 4 | 949 | 73 | ||
| NM_128708.5 | NP_180709.3 | 3 - 4 | 799 | 73 | ||
Set Membership: |
|