Search
Search

XIF| Gene Symbol: XIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336351.1 | NP_001318329.1 | 40 - 41 | 5248 | 73 | |
| NM_001336352.1 | NP_001323600.1 | 38 - 39 | 4538 | 73 | ||
| NM_001336353.1 | NP_001323601.1 | 33 - 34 | 3764 | 73 | ||
| NM_001336354.1 | NP_001323599.1 | 36 - 37 | 4329 | 73 | ||
| NM_128748.3 | NP_180749.2 | 38 - 39 | 4535 | 73 | ||
Set Membership: |
|