Search
Search

IAA13| Gene Symbol: IAA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124962.1 | NP_001118434.1 | 3 - 4 | 960 | 67 | |
| NM_128891.4 | NP_180889.1 | 3 - 4 | 1025 | 67 | ||
| NM_179874.2 | NP_850205.1 | 3 - 4 | 908 | 67 | ||
| GenBank mRNA | AF332399.1 | - | 3 - 4 | 693 | 67 | |
| AK227387.1 | - | 3 - 4 | 873 | 67 | ||
| AY085194.1 | - | 3 - 4 | 934 | 67 | ||
| BT005874.1 | - | 3 - 4 | 696 | 67 | ||
| U18415.1 | - | 3 - 4 | 909 | 67 | ||
Set Membership: |
|