Search
Search

RD20| Gene Symbol: RD20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036395.1 | NP_001031472.1 | 1 - 2 | 198 | 68 | |
| NM_128898.4 | NP_180896.1 | 1 - 2 | 282 | 68 | ||
| GenBank mRNA | AB039924.1 | - | 1 - 2 | 172 | 68 | |
| AK317039.1 | - | 1 - 2 | 195 | 68 | ||
| AY062661.1 | - | 1 - 2 | 198 | 68 | ||
| AY063964.1 | - | 1 - 2 | 193 | 68 | ||
| AY093321.1 | - | 1 - 2 | 101 | 68 | ||
| AY096405.1 | - | 1 - 2 | 101 | 68 | ||
Set Membership: |
|