Search
Search

WDR55| Gene Symbol: WDR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128977.4 | NP_565782.1 | 2 - 3 | 509 | 70 | |
| NM_201867.2 | NP_973596.1 | 2 - 3 | 149 | 70 | ||
| GenBank mRNA | AK117481.1 | - | 2 - 3 | 176 | 70 | |
| AY087666.1 | - | 2 - 3 | 176 | 70 | ||
| BT005438.1 | - | 2 - 3 | 126 | 70 | ||
Set Membership: |
|