Search
Search

AT1G51610| Gene Symbol: AT1G51610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104039.4 | NP_564594.1 | 9 - 10 | 1446 | 100 | |
| GenBank mRNA | AY150383.1 | - | 9 - 10 | 1035 | 100 | |
Set Membership: |
|