Search
Search

B ALPHA| Gene Symbol: B ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198270.1 | NP_001185199.1 | 13 - 14 | 1579 | 85 | |
| NM_001333494.1 | NP_001322512.1 | 11 - 12 | 3478 | 85 | ||
| NM_001333495.1 | NP_001322511.1 | 13 - 14 | 1600 | 85 | ||
| NM_104047.4 | NP_564595.1 | 12 - 13 | 3207 | 85 | ||
| NM_202274.2 | NP_974003.1 | 11 - 12 | 1501 | 85 | ||
| GenBank mRNA | AK230310.1 | - | 12 - 13 | 2987 | 85 | |
| AY120756.1 | - | 11 - 12 | 1502 | 85 | ||
| BT000109.1 | - | 11 - 12 | 1260 | 85 | ||
Set Membership: |
|