Search
Search

UBP6| Gene Symbol: UBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104049.4 | NP_564596.1 | 2 - 3 | 458 | 70 | |
| GenBank mRNA | AY050817.1 | - | 2 - 3 | 222 | 70 | |
| AY084730.1 | - | 2 - 3 | 226 | 70 | ||
| AY114081.1 | - | 1 - 2 | 157 | 70 | ||
| BT000658.1 | - | 2 - 3 | 227 | 70 | ||
Set Membership: |
|