Search
Search

LHCA2| Gene Symbol: LHCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116012.5 | NP_191706.2 | 3 - 4 | 973 | 116 | |
| GenBank mRNA | AF134120.1 | - | 3 - 4 | 789 | 116 | |
| AY054670.1 | - | 3 - 4 | 791 | 116 | ||
| AY065429.1 | - | 3 - 4 | 818 | 116 | ||
| AY072483.1 | - | 3 - 4 | 751 | 116 | ||
| BT002070.1 | - | 3 - 4 | 791 | 116 | ||
Set Membership: |
|