Search
Search

ARF18| Gene Symbol: ARF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340129.1 | NP_001327870.1 | 10 - 11 | 1654 | 69 | |
| NM_116048.3 | NP_567119.1 | 11 - 12 | 1762 | 69 | ||
| GenBank mRNA | AF334717.1 | - | 11 - 12 | 1480 | 69 | |
| AY059746.1 | - | 11 - 12 | 1700 | 69 | ||
| AY070463.1 | - | 11 - 12 | 1765 | 69 | ||
| AY091392.1 | - | 11 - 12 | 1480 | 69 | ||
Set Membership: |
|