Search
Search

ARF18| Gene Symbol: ARF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340129.1 | NP_001327870.1 | 7 - 8 | 989 | 114 | |
| NM_116048.3 | NP_567119.1 | 8 - 9 | 1097 | 114 | ||
| GenBank mRNA | AF334717.1 | - | 8 - 9 | 815 | 114 | |
| AY059746.1 | - | 8 - 9 | 1035 | 114 | ||
| AY070463.1 | - | 8 - 9 | 1100 | 114 | ||
| AY091392.1 | - | 8 - 9 | 815 | 114 | ||
Set Membership: |
|