Search
Search

GID1B| Gene Symbol: GID1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116166.5 | NP_191860.1 | 1 - 2 | 512 | 70 | |
| GenBank mRNA | AK228182.1 | - | 1 - 2 | 330 | 70 | |
| BT028997.1 | - | 1 - 2 | 38 | 70 | ||
Set Membership: |
|