Search
Search

PARP2| Gene Symbol: PARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340373.1 | NP_001329308.1 | 13 - 14 | 1583 | 103 | |
| NM_001340374.1 | NP_001329307.1 | 12 - 13 | 1650 | 103 | ||
| NM_116472.4 | NP_192148.2 | 13 - 14 | 1757 | 103 | ||
Set Membership: |
|