Search
Search

EHD2| Gene Symbol: EHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116790.3 | NP_567299.2 | 13 - 14 | 1301 | 65 | |
| NM_202788.1 | NP_974517.1 | 13 - 14 | 1250 | 65 | ||
| GenBank mRNA | BT033036.1 | - | 13 - 14 | 1182 | 65 | |
Set Membership: |
|