Search
Search

KNAT1| Gene Symbol: KNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116884.4 | NP_192555.1 | 2 - 3 | 1141 | 95 | |
| GenBank mRNA | AY080834.1 | - | 2 - 3 | 697 | 95 | |
| AY113982.1 | - | 2 - 3 | 592 | 95 | ||
Set Membership: |
|