Search
Search

TCP3| Gene Symbol: TCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104201.3 | NP_564624.2 | 1 - 2 | 1523 | 102 | |
| GenBank mRNA | AY099678.1 | - | 1 - 2 | 1503 | 102 | |
| BT000266.1 | - | 1 - 2 | 1233 | 102 | ||
Set Membership: |
|