Search
Search

LOX1| Gene Symbol: LOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104376.3 | NP_175900.1 | 6 - 7 | 1706 | 81 | |
| GenBank mRNA | AY093104.1 | - | 6 - 7 | 1650 | 81 | |
| BT010358.1 | - | 6 - 7 | 1562 | 81 | ||
| L04637.1 | - | 6 - 7 | 1642 | 81 | ||
Set Membership: |
|