Search
Search

AAE18| Gene Symbol: AAE18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084254.1 | NP_001077723.1 | 5 - 6 | 904 | 65 | |
| NM_104408.5 | NP_175929.3 | 5 - 6 | 1167 | 65 | ||
| GenBank mRNA | AY250845.1 | - | 5 - 6 | 892 | 65 | |
Set Membership: |
|