Search
Search

GolS2| Gene Symbol: GolS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104537.3 | NP_176053.1 | 1 - 2 | 414 | 121 | |
| GenBank mRNA | AB062849.1 | - | 1 - 2 | 410 | 121 | |
| AY050410.1 | - | 1 - 2 | 414 | 121 | ||
| AY058238.1 | - | 1 - 2 | 331 | 121 | ||
| KJ138960.1 | - | 1 - 2 | 331 | 121 | ||
Set Membership: |
|