Search
Search

RXF12| Gene Symbol: RXF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333829.1 | NP_001323263.1 | 3 - 4 | 773 | 84 | |
| NM_104617.4 | NP_176133.1 | 3 - 4 | 470 | 84 | ||
| GenBank mRNA | AB008015.1 | - | 3 - 4 | 452 | 84 | |
Set Membership: |
|