Search
Search

XF1| Gene Symbol: XF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104624.4 | NP_564734.1 | 1 - 2 | 382 | 82 | |
| GenBank mRNA | AB008021.1 | - | 1 - 2 | 276 | 82 | |
| AY099643.1 | - | 1 - 2 | 292 | 82 | ||
| BT000239.1 | - | 1 - 2 | 248 | 82 | ||
Set Membership: |
|