Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: E1 ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333870.1 | NP_001321393.1 | 5 - 6 | 1131 | 135 | |
NM_104682.3 | NP_176198.1 | 5 - 6 | 1293 | 135 | ||
GenBank mRNA | AY087667.1 | - | 5 - 6 | 1118 | 135 | |
BT000974.1 | - | 5 - 6 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |