Search
Search

ABC4| Gene Symbol: ABC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124046.2 | NP_001117518.1 | 10 - 11 | 1415 | 119 | |
| NM_104743.3 | NP_176259.2 | 10 - 11 | 1407 | 119 | ||
| GenBank mRNA | AK227267.1 | - | 10 - 11 | 1338 | 119 | |
Set Membership: |
|