Search
Search

MLO6| Gene Symbol: MLO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198352.1 | NP_001185281.1 | 9 - 10 | 981 | 106 | |
| NM_001198354.1 | NP_001185283.1 | 9 - 10 | 983 | 106 | ||
| NM_001333983.1 | NP_001322619.1 | 9 - 10 | 1201 | 106 | ||
| NM_001333984.1 | NP_001319285.1 | 9 - 10 | 1146 | 106 | ||
| NM_001333985.1 | NP_001322617.1 | 9 - 10 | 1225 | 106 | ||
| NM_001333986.1 | NP_001322615.1 | 9 - 10 | 1225 | 106 | ||
| NM_001333987.1 | NP_001322616.1 | 9 - 10 | 1225 | 106 | ||
| NM_001333988.1 | NP_001322614.1 | 9 - 10 | 1225 | 106 | ||
| NM_001333989.1 | NP_001322618.1 | 9 - 10 | 1201 | 106 | ||
| NM_104836.2 | NP_176350.1 | 9 - 10 | 1225 | 106 | ||
| GenBank mRNA | AF369567.2 | - | 9 - 10 | 980 | 106 | |
Set Membership: |
|