Search
Search

NRPD1A| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 5 - 6 | 430 | 97 | |
| NM_001334061.1 | NP_001320709.1 | 4 - 5 | 620 | 97 | ||
| NM_001334062.1 | NP_001320708.1 | 5 - 6 | 430 | 97 | ||
| NM_104980.4 | NP_176490.2 | 4 - 5 | 595 | 97 | ||
| GenBank mRNA | AY826515.1 | - | 3 - 4 | 321 | 97 | |
| DQ020657.1 | - | 4 - 5 | 595 | 97 | ||
Set Membership: |
|