Search
Search

ABCC2| Gene Symbol: ABCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202746.1 | NP_001189675.1 | 23 - 24 | 3541 | 91 | |
| NM_001336522.1 | NP_001324371.1 | 17 - 18 | 2514 | 91 | ||
| NM_129020.3 | NP_181013.1 | 22 - 23 | 3806 | 91 | ||
Set Membership: |
|