Search
Search

RPA| Gene Symbol: RPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129074.3 | NP_565801.1 | 3 - 4 | 759 | 92 | |
| NM_201874.2 | NP_973603.1 | 3 - 4 | 759 | 92 | ||
| GenBank mRNA | AK229368.1 | - | 3 - 4 | 707 | 92 | |
Set Membership: |
|