Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129074.3 | NP_565801.1 | 6 - 7 | 1247 | 70 | |
NM_201874.2 | NP_973603.1 | 6 - 7 | 1237 | 70 | ||
GenBank mRNA | AK229368.1 | - | 6 - 7 | 1195 | 70 | |
AY085599.1 | - | 6 - 7 | 1247 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |