Search
Search

TET10| Gene Symbol: TET10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036145.3 | NP_001031222.1 | 8 - 9 | 851 | 80 | |
| NM_001334076.1 | NP_001320677.1 | 8 - 9 | 870 | 80 | ||
| NM_001334077.1 | NP_001320676.1 | 9 - 10 | 769 | 80 | ||
| NM_105005.5 | NP_176515.3 | 8 - 9 | 851 | 80 | ||
| NM_202348.3 | NP_974077.1 | 8 - 9 | 851 | 80 | ||
| GenBank mRNA | BT011569.1 | - | 8 - 9 | 80 | ||
| BT012253.1 | - | 8 - 9 | 80 | |||
Set Membership: |
|