Search
Search

ABCF3| Gene Symbol: ABCF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105130.4 | NP_176636.1 | 12 - 13 | 1702 | 118 | |
| GenBank mRNA | BT002015.1 | - | 12 - 13 | 1644 | 118 | |
| BT010544.1 | - | 12 - 13 | 1602 | 118 | ||
Set Membership: |
|