Search
Search

ABCF3| Gene Symbol: ABCF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105130.4 | NP_176636.1 | 13 - 14 | 1801 | 73 | |
| GenBank mRNA | BT002015.1 | - | 13 - 14 | 1743 | 73 | |
| BT010544.1 | - | 13 - 14 | 1701 | 73 | ||
Set Membership: |
|