Search
Search

AT1G65150| Gene Symbol: AT1G65150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105188.2 | NP_176694.1 | 5 - 6 | 829 | 63 | |
| NM_202363.3 | NP_974092.1 | 5 - 6 | 876 | 63 | ||
| GenBank mRNA | AY128307.1 | - | 5 - 6 | 835 | 63 | |
| BT001166.1 | - | 4 - 5 | 734 | 63 | ||
Set Membership: |
|