Search
Search

UNE16| Gene Symbol: UNE16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036549.2 | NP_001031626.1 | 1 - 2 | 337 | 99 | |
| NM_117438.4 | NP_567408.1 | 1 - 2 | 397 | 99 | ||
| GenBank mRNA | AK226443.1 | - | 1 - 2 | 255 | 99 | |
| AY087771.1 | - | 1 - 2 | 322 | 99 | ||
Set Membership: |
|