Search
Search

XTH15| Gene Symbol: XTH15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117490.4 | NP_193149.2 | 2 - 3 | 663 | 100 | |
| GenBank mRNA | AY045865.1 | - | 2 - 3 | 552 | 100 | |
| AY087282.1 | - | 2 - 3 | 552 | 100 | ||
| BT020422.1 | - | 2 - 3 | 467 | 100 | ||
| U43489.1 | - | 2 - 3 | 467 | 100 | ||
Set Membership: |
|