Search
Search

CKI1| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340918.1 | NP_001328596.1 | 8 - 9 | 977 | 82 | |
| NM_117511.5 | NP_193170.1 | 8 - 9 | 977 | 82 | ||
| GenBank mRNA | AF250343.1 | - | 8 - 9 | 801 | 82 | |
| AY052364.1 | - | 8 - 9 | 947 | 82 | ||
| AY060528.1 | - | 8 - 9 | 647 | 82 | ||
| AY943850.1 | - | 8 - 9 | 647 | 82 | ||
| X78818.1 | - | 818 | 82 | |||
Set Membership: |
|