Search
Search

KMS1| Gene Symbol: KMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117581.5 | NP_567450.1 | 5 - 6 | 1294 | 111 | |
| NM_202822.1 | NP_974551.1 | 5 - 6 | 1151 | 111 | ||
| NM_202823.2 | NP_974552.1 | 5 - 6 | 1193 | 111 | ||
| GenBank mRNA | BT015780.1 | - | 5 - 6 | 802 | 111 | |
Set Membership: |
|