Search
Search

CYP97B3| Gene Symbol: CYP97B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117600.6 | NP_193247.2 | 1 - 2 | 282 | 81 | |
| GenBank mRNA | AY062675.1 | - | 1 - 2 | 190 | 81 | |
| BT002582.1 | - | 1 - 2 | 157 | 81 | ||
Set Membership: |
|