Search
Search

BAM5| Gene Symbol: BAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340998.1 | NP_001329368.1 | 4 - 5 | 1291 | 90 | |
| NM_001340999.1 | NP_001329367.1 | 4 - 5 | 1464 | 90 | ||
| NM_117609.3 | NP_567460.1 | 4 - 5 | 1066 | 90 | ||
| NM_179058.1 | NP_849389.1 | 4 - 5 | 1010 | 90 | ||
| GenBank mRNA | AF424573.1 | - | 4 - 5 | 1005 | 90 | |
| AK226353.1 | - | 4 - 5 | 1008 | 90 | ||
| AK316869.1 | - | 4 - 5 | 1006 | 90 | ||
| AY142024.1 | - | 4 - 5 | 897 | 90 | ||
| M73467.1 | - | 986 | 90 | |||
Set Membership: |
|