Search
Search

PPDK| Gene Symbol: PPDK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036570.1 | NP_001031647.1 | 6 - 7 | 833 | 65 | |
| NM_001084926.1 | NP_001078395.1 | 5 - 6 | 770 | 65 | ||
| NM_001084927.2 | NP_001078396.1 | 4 - 5 | 822 | 65 | ||
| NM_001125515.1 | NP_001118987.1 | 4 - 5 | 715 | 65 | ||
| NM_001341051.1 | NP_001328869.1 | 4 - 5 | 941 | 65 | ||
| NM_117643.4 | NP_193288.2 | 3 - 4 | 571 | 65 | ||
| NM_179060.4 | NP_849391.2 | 5 - 6 | 809 | 65 | ||
| GenBank mRNA | AK317187.1 | - | 5 - 6 | 798 | 65 | |
| AK317578.1 | - | 3 - 4 | 489 | 65 | ||
Set Membership: |
|