Search
Search

MAA3| Gene Symbol: MAA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341063.1 | NP_001329006.1 | 13 - 14 | 1800 | 77 | |
| NM_001341064.1 | NP_001329005.1 | 13 - 14 | 1556 | 77 | ||
| NM_001341065.1 | NP_001329007.1 | 11 - 12 | 1271 | 77 | ||
| NM_117648.8 | NP_193292.3 | 14 - 15 | 1593 | 77 | ||
| GenBank mRNA | EU915246.1 | - | 14 - 15 | 1593 | 77 | |
Set Membership: |
|