Search
Search

CT-BMY| Gene Symbol: CT-BMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117813.3 | NP_567523.1 | 2 - 3 | 895 | 68 | |
| GenBank mRNA | AJ250341.1 | - | 2 - 3 | 790 | 68 | |
| AY087592.1 | - | 2 - 3 | 827 | 68 | ||
Set Membership: |
|